Geraniol 95%

Supplier: Ambeed
Danger

A107131-100G A107131-25G A107131-500G A107131-1000G
76796-596EA 32 USD
76796-596 76796-598 76796-600 77599-554
Geraniol 95%
Geraniol

Caution: for Research Use Only

Formula: C₁₀H₁₈O
MDL Number: MFCD00002917
CAS Number: 106-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR