2-Amino-3-(naphthalen-1-yl)propanoic acid 95%
Supplier: Ambeed
|
Warning
|
A104917-10G
A104917-1G
A104917-250MG
A104917-5G
76796-442EA
53
USD
76796-442
76796-444
76796-446
76796-448
2-Amino-3-(naphthalen-1-yl)propanoic acid 95%
2-Amino-3-(naphthalen-1-yl)propanoic acid
Caution: for Research Use Only
Formula:
C₁₃H₁₃NO₂ |
MDL Number:
MFCD00067507 CAS Number: 28095-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...