HTD Equilibrium Dialysis Device

Supplier: HTDIALYSIS LLC MS
1006 1009 1101 1103 1104 1105 1135 1150 1151
104018-890EA 7406.77 USD
104018-890 104018-868 104018-874 104018-898 104018-864 104018-882 104018-894 104018-896 104018-892
HTD Equilibrium Dialysis Device
Tubing Dialysis Tubing
HTD reusable equilibrium dialysis device combines industry-leading accuracy and precision with an automation compatible design for a cost efficient, scalable solution.

  • Simultaneously process 1 to 96 assays, each with a 30 to 150 µl capacity to conserve valuable reagents
  • Compatible with a wide range of dialysis membrane pore sizes
  • Ready in minutes, achieves equilibrium in 4 to 6 hours on average
  • Automatable with standard robotic workstations and 96-well pipettors
  • Ready access to both the sample and dialysate sides of the membrane from the top of the device
  • Minimal non-specific binding due to 100% Teflon block construction
  • Reusable device is cost-effective, eco-friendly, and scalable
  • Compatible with a wide range of dialysis membrane pore sizes: 1 kDa, 3.5 kDa, 6 to 8 kDa, 10 kDa, 12 to 14 kDa, 25 kDa, 50 kDa

The HTD96B micro equilibrium device was developed at a Global Pharmaceutical Research laboratory to assess serum protein binding, microsomal protein binding, and tissue specific binding of drug candidates, conserving sample quantities, and achieving rapid equilibrium.

Reusable device combines industry-leading accuracy and precision with an automation compatible design for a scalable, cost-efficient, and eco-friendly solution.

Applications:
- Serum/plasma protein binding
- Microsome protein binding
- Relative binding of drug to specific tissues
- Receptor binding
- T3/T4 free fraction binding (hyperthyroidism) determinations

The 96C model has a shorter diffusion path with a 25 to 75 µl capacity and achieves equilibrium in 2 to 4 hours on average.

Additional specifications:
Dimensions: 6.7×4.6×1.5"
Weight: 1.2 lbs.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR