Alimemazine Tartrate 98%

Supplier: Ambeed
Danger

Synonyms: Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate, Alimemazine Hemitartrate

A954362-100MG A954362-10MG A954362-1MG A954362-50MG
76792-214EA 37 USD
76792-214 76792-216 76792-218 76792-220
Alimemazine Tartrate 98%
Alimemazine Tartrate

Caution: for Research Use Only

Formula: C₃₆H₄₄N₄S₂·C₄H₆O₆
MW: 746.98 g/mol
MDL Number: MFCD00242594
CAS Number: 4330-99-8
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR