2-Bromo-3,4-difluoroaniline 97%

Supplier: Ambeed
Warning

A954208-100G A954208-10G A954208-1G A954208-25G A954208-5G
76792-204EA 287 USD
76792-204 76792-206 76792-208 76792-210 76792-212
2-Bromo-3,4-difluoroaniline 97%
2-Bromo-3,4-difluoroaniline

Caution: for Research Use Only

Formula: C₆H₄BrF₂N
MW: 208.00 g/mol
MDL Number: MFCD11035924
CAS Number: 1092349-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR