2,4,5-Trichloro-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A890578-5G A890578-1G A890578-100MG A890578-250MG
76792-038EA 88 USD
76792-038 77603-226 77873-339 77873-340
2,4,5-Trichloro-7H-pyrrolo[2,3-d]pyrimidine 98%
2,4,5-Trichloro-7H-pyrrolo[2,3-d]pyrimidine

Caution: for Research Use Only

Formula: C₆H₂Cl₃N₃
MDL Number: MFCD13193620
CAS Number: 1053228-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR