Xanthone 98%

Supplier: Ambeed
Danger

Synonyms: Xanthen-9-one, 9H-Xanthen-9-one

A837363-250MG A837363-25G A837363-5G A837363-10G
76791-846EA 5 USD
76791-846 76791-848 76791-850 77603-128
Xanthone 98%
Xanthone

Caution: for Research Use Only

Formula: C₁₃H₈O₂
MW: 196.21 g/mol
Boiling Pt: 349
Melting Pt: 174
MDL Number: MFCD00005060
CAS Number: 90-47-1
UN: 2811
ADR: 6.1 Ⅲ
Merck Index: 12,10196

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR