2,3',4,5'-Tetramethoxystilbene 98%

Supplier: Ambeed
Warning

A779383-100MG A779383-10MG A779383-1MG A779383-25MG A779383-50MG A779383-5MG A779383-1G A779383-5G A779383-250MG
76791-554EA 61 USD
76791-554 76791-556 76791-558 76791-560 76791-562 76791-564 77670-088 77670-090 77873-236
2,3',4,5'-Tetramethoxystilbene 98%
2,3',4,5'-Tetramethoxystilbene

Caution: for Research Use Only

Formula: C₁₈H₂₀O₄
MDL Number: MFCD03428621
CAS Number: 24144-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR