4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-(4-Aminobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene, 4-Aminobenzo-15-crown 5-Ether, 4'-Aminobenzo-15-crown 5-Ether

A570923-100MG A570923-1G A570923-250MG A570923-5G
76790-348EA 11 USD
76790-348 76790-350 76790-352 77872-951
4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%
4'-Aminobenzo-15-crown-5 (4'-Aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane)

Caution: for Research Use Only

Formula: C₁₄H₂₁NO₅
MDL Number: MFCD00068686
CAS Number: 60835-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR