Triphenyl phosphate 99%

Supplier: Ambeed
Danger

A548113-1000G A548113-500G
76790-246EA 6.37 USD
76790-246 77602-316 77602-318
Triphenyl phosphate 99%
Triphenyl phosphate

Caution: for Research Use Only

Formula: C₁₈H₁₅O₄P
MDL Number: MFCD00003031
CAS Number: 115-86-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR