(3-Acetamidophenyl)boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide, 3-Acetamidophenylboronic acid Pinacol ester, 3-Acetamidophenylboronic acid, pinacol estet, 2-(3-Acetamidophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide

A500419-1G A500419-250MG A500419-25G A500419-5G
76789-924EA 28 USD
76789-924 76789-926 76789-928 76789-930
(3-Acetamidophenyl)boronic acid pinacol ester 98%
(3-Acetamidophenyl)boronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₄H₂₀BNO₃
MDL Number: MFCD03789261
CAS Number: 480424-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR