Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Al(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)aluminum
A471644-1G
A471644-25G
A471644-5G
76789-572EA
22
USD
76789-572
76789-574
76789-576
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium 98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium
Caution: for Research Use Only
Formula:
C₃₃H₅₇AlO₆ |
MDL Number:
MFCD00058918 CAS Number: 14319-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...