D-α-Tocopherol 98%

Supplier: Ambeed
Warning

Synonyms: (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol

A443508-1G A443508-25G A443508-5G A443508-500G
76789-288EA 6 USD
76789-288 76789-290 76789-292 77668-980
D-α-Tocopherol 98%
D-α-Tocopherol

Caution: for Research Use Only

Formula: C₂₉H₅₀O₂
MW: 430.71 g/mol
MDL Number: MFCD00072045
CAS Number: 59-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR