Eriodictyol 98%

Supplier: Ambeed
Warning

A404191-1MG A404191-100MG
76788-970EA 18.04 USD
76788-970 77668-844
Eriodictyol 98%
Eriodictyol

Caution: for Research Use Only

Formula: C₁₅H₁₂O₆
MDL Number: MFCD00135890
CAS Number: 552-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR