2,2-Diphenylglycine 95%
Supplier: Ambeed
|
Warning
|
A357964-100G
A357964-10G
A357964-1G
A357964-250MG
A357964-25G
A357964-5G
76788-518EA
483
USD
76788-518
76788-520
76788-522
76788-524
76788-526
76788-528
2,2-Diphenylglycine 95%
2,2-Diphenylglycine
Caution: for Research Use Only
Formula:
C₁₄H₁₃NO₂ |
MDL Number:
MFCD00008048 CAS Number: 3060-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...