2,4,6-Triphenylpyridine 98%

Supplier: Ambeed
Danger

A347756-1G A347756-250MG A347756-25G A347756-5G
76788-348EA 26 USD
76788-348 76788-350 76788-352 76788-354
2,4,6-Triphenylpyridine 98%
2,4,6-Triphenylpyridine

Caution: for Research Use Only

Formula: C₂₃H₁₇N
MW: 307.40 g/mol
MDL Number: MFCD00014630
CAS Number: 580-35-8
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR