Vinpocetine 98%

Supplier: Ambeed
Warning

Synonyms: (41S,13aS)-Ethyl 13a-ethyl-2,3,41,5,6,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate

A305598-100G A305598-100MG A305598-10G A305598-1G A305598-250MG A305598-25G A305598-5G
76787-828EA 443 USD
76787-828 76787-830 76787-832 76787-834 76787-836 76787-838 76787-840
Vinpocetine 98%
Vinpocetine

Caution: for Research Use Only

Formula: C₂₂H₂₆N₂O₂
MDL Number: MFCD00211233
CAS Number: 42971-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR