Sulfaguanidine 98% (< 8% H₂O)

Supplier: Ambeed
Warning

A293011-25G A293011-100MG A293011-500G A293011-1000G A293011-100G
76787-582EA 8 USD
76787-582 77668-296 77668-298 77872-375 77872-376
Sulfaguanidine 98% (< 8% H₂O)
Sulfaguanidine

Caution: for Research Use Only

Formula: C₇H₁₀N₄O₂S
MDL Number: MFCD00038136
CAS Number: 57-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR