9-(Chloromethyl)anthracene 95%

Supplier: Ambeed
Danger

A290315-10G A290315-1G A290315-25G A290315-5G
76787-532EA 38 USD
76787-532 76787-534 76787-536 76787-538
9-(Chloromethyl)anthracene 95%
9-(Chloromethyl)anthracene

Caution: for Research Use Only

Formula: C₁₅H₁₁Cl
MDL Number: MFCD00001263
CAS Number: 24463-19-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR