(6-Aminopyridin-3-yl)boronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Aminopyridine-3-boronic acid, (6-aminopyridin-3-yl)boronic acid
A276340-10G
A276340-1G
A276340-5G
76787-280EA
49
USD
76787-280
76787-282
76787-284
(6-Aminopyridin-3-yl)boronic acid 95%
(6-Aminopyridin-3-yl)boronic acid
Caution: for Research Use Only
Formula:
C₅H₇BN₂O₂ MW: 137.93 g/mol |
MDL Number:
MFCD09907980 CAS Number: 851524-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...