Tris(2,2,2-trifluoroethyl) Phosphate 98%

Supplier: Ambeed
Danger

Synonyms: Tris(2,2,2-trifluoroethyl)phosphate, Phosphoric acid tris(2,2,2-trifluoroethyl) ester

A269962-25G A269962-100G
76787-172EA 24 USD
76787-172 77872-313
Tris(2,2,2-trifluoroethyl) Phosphate 98%
Tris(2,2,2-trifluoroethyl) Phosphate

Caution: for Research Use Only

Formula: C₆H₆F₉O₄P
MDL Number: MFCD00233117
CAS Number: 358-63-4
UN: 3272
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR