3-Amino-5-cyclobutyl-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
A258874-1G
A258874-5G
76786-936EA
447.02
USD
76786-936
76786-934
3-Amino-5-cyclobutyl-1H-pyrazole 97%
3-Amino-5-cyclobutyl-1H-pyrazole
Caution: for Research Use Only
Formula:
C₇H₁₁N₃ |
MDL Number:
MFCD08061045 CAS Number: 326827-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...