(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-(hydroxymethyl)oxazolidione 98%

Supplier: Ambeed
Warning

Synonyms: (5R)-3-(3-Fluoro-4-(4-morpholinyl)phenyl)-5-hydroxymethyl-2-oxazolidione

A246816-100G A246816-1G A246816-250MG A246816-25G A246816-5G
76786-702EA 265 USD
76786-702 76786-704 76786-706 76786-708 76786-710
(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-(hydroxymethyl)oxazolidione 98%
(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-(hydroxymethyl)oxazolidione

Caution: for Research Use Only

Formula: C₁₄H₁₇FN₂O₄
MDL Number: MFCD06658241
CAS Number: 168828-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR