4-Benzyloxy-2-formylphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-(Benzyloxy)-2-formylphenyl)boronic acid

A235702-1G A235702-25G A235702-5G A235702-250MG
76786-398EA 36.08 USD
76786-398 76786-400 76786-402 77600-894
4-Benzyloxy-2-formylphenylboronic acid 95%
4-Benzyloxy-2-formylphenylboronic acid

Caution: for Research Use Only

Formula: C₁₄H₁₃BO₄
MW: 256.0616 g/mol
MDL Number: MFCD02179486
CAS Number: 139962-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR