Xylitol 98+%

Supplier: Ambeed
Warning

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

A221974-1000G A221974-5G A221974-25G A221974-500G A221974-100G
76786-030EA 8.49 USD
76786-030 76786-028 76786-036 76786-034 76786-032
Xylitol 98+%
Xylitol

Caution: for Research Use Only

Formula: C₅H₁₂O₅
MW: 152.15 g/mol
MDL Number: MFCD00064292
CAS Number: 87-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR