YC-1 98%

Supplier: Ambeed
Warning

A174113-1MG A174113-50MG A174113-5MG A174113-100MG A174113-250MG
76784-336EA 13 USD
76784-336 76784-338 76784-340 77667-430 77871-869
YC-1 98%
YC-1

Caution: for Research Use Only

Formula: C₁₉H₁₆N₂O₂
MDL Number: MFCD06407798
CAS Number: 170632-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR