Succinimide 97%

Supplier: Ambeed
Warning

Synonyms: Pyrrolidine-2,5-dione

A168239-1000G A168239-100G A168239-25G A168239-500G
76784-130EA 27 USD
76784-130 76784-132 76784-134 76784-136
Succinimide 97%
Succinimide

Caution: for Research Use Only

Formula: C₄H₅NO₂
MDL Number: MFCD00005495
CAS Number: 123-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR