5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one Hydrochloride

A153551-1G A153551-5G A153551-25G
76783-484EA 11 USD
76783-484 76783-486 77600-184
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride 95%
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride

Caution: for Research Use Only

Formula: C₇H₁₀ClNOS
MDL Number: MFCD11111130
CAS Number: 115473-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR