Tetrathiafulvalene 98%

Supplier: Ambeed
Warning

A149342-100MG A149342-1G A149342-250MG A149342-5G
76783-296EA 13 USD
76783-296 76783-298 76783-300 76783-302
Tetrathiafulvalene 98%
Tetrathiafulvalene

Caution: for Research Use Only

Formula: C₆H₄S₄
MDL Number: MFCD00005492
CAS Number: 31366-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR