Tetraethylene glycol dimethyl ether 97%

Supplier: Ambeed
Warning

A139639-1000G A139639-100G A139639-500G
76782-880EA 59.42 USD
76782-880 76782-882 76782-884
Tetraethylene glycol dimethyl ether 97%
Tetraethylene glycol dimethyl ether

Caution: for Research Use Only

Formula: C₁₀H₂₂O₅
MDL Number: MFCD00008505
CAS Number: 143-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR