2,3-Bis(amino((2-aminophenyl)thio)methylene)succinonitrile compound with ethanol (1:1) 98%

Supplier: Ambeed
Warning

A132258-100MG A132258-10MG A132258-1MG A132258-50MG A132258-5MG A132258-1G A132258-250MG
76782-416EA 97.01 USD
76782-416 76782-418 76782-420 76782-422 76782-424 77871-621 77871-622
2,3-Bis(amino((2-aminophenyl)thio)methylene)succinonitrile compound with ethanol (1:1) 98%
2,3-Bis(amino((2-aminophenyl)thio)methylene)succinonitrile compound with ethanol (1:1)

Caution: for Research Use Only

Formula: C₂₀H₂₂N₆OS₂
MDL Number: MFCD22628895
CAS Number: 1173097-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR