Hair and Dander Vacuum for Shavers
Supplier: Stoelting
51490
100229-642EA 5878.08 USD
100229-642
Hair and Dander Vacuum for Shavers
Vacuum Cleaners
The Hair and Dander vacuum attaches to most Oster and Andis clippers (not included), including the Andis AG and the Oster A5. Hair and dander are vacuumed away as soon as they are cut, then collected in a small portable vacuum cleaner.
The vacuum cleaner is equipped with a paper bag and a cloth filter backed by a HEPA filter. This combination is 99.97% efficient to 0.3 microns, without excessive filter replacement costs. The exhaust to the room is only clean air.
Accessories information: The paper bag filters have a capacity of 8.3 dry quarts.
Delivery information: The system comes complete with the vacuum cleaner on a cart, the clipper attachment head, and 3 paper bag filters.