5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride 95%

Supplier: Ambeed
Warning

Synonyms: 6-(2,5-Dioxotetrahydrofuran-3-yl)-4-methyl-7,7a-dihydroisobenzofuran-1,3(3aH,6H)-dione

A896652-1G A896652-5G
76776-926EA 9 USD
76776-926 76776-928
5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride 95%
5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride

Caution: for Research Use Only

Formula: C₁₃H₁₂O₆
MW: 264.23 g/mol
MDL Number: MFCD00039141
CAS Number: 73003-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR