(4-Bromophenyl)acetyl chloride 97%
Supplier: Ambeed
|
Danger
|
A887814-1G
A887814-25G
A887814-5G
76776-914EA
20
USD
76776-914
76776-916
76776-918
(4-Bromophenyl)acetyl chloride 97%
(4-Bromophenyl)acetyl chloride
Caution: for Research Use Only
Formula:
C₈H₆BrClO |
MDL Number:
MFCD00126842 CAS Number: 37859-24-8 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...