Sodium octanoate 99%

Supplier: Ambeed
Warning

Synonyms: Caprylic acid sodium acidOctanoic acid sodium acidSodium n-OctanoateSodium caprylate

A869967-1000G A869967-25G A869967-500G A869967-100G
76776-832EA 25.47 USD
76776-832 76776-826 76776-828 76776-830
Sodium octanoate 99%
Sodium octanoate

Caution: for Research Use Only

Formula: C₈H₁₅NaO₂
MW: 166.2 g/mol
MDL Number: MFCD00058511
CAS Number: 1984-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR