4-Amino-N-methylbenzenemethanesulfonamide 98%

Supplier: Ambeed
Danger

Synonyms: 4-Amino-N-methyl-alpha-toluenesulfonamide, 4-Amino-N-methyl-α-toluenesulphonamide, 4-Amino-N-methyl-α-toluenesulfonamide, 1-(4-Aminophenyl)-N-methylmethanesulphonamide, 4-Amino-N-methylbenzenemethanesulphonamide, 4-Amino-N-methyl-alpha-toluenesulphonamide, 4-Amino-N-methylbenzenemethanesulfonamide, 1-(4-Aminophenyl)-N-methylmethanesulfonamide

A519346-250MG A519346-25G
76774-636EA 10 USD
76774-636 77878-305
4-Amino-N-methylbenzenemethanesulfonamide 98%
4-Amino-N-methylbenzenemethanesulfonamide

Caution: for Research Use Only

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD00671697
CAS Number: 109903-35-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR