5-TAMRA 98%

Supplier: Ambeed
Warning

Synonyms: 5-CarboxytetramethylrhodamineTAMRA, 5-isomer2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate

A505764-100MG A505764-5G A505764-1G A505764-250MG A505764-10MG A505764-1MG A505764-5MG A505764-25MG
76774-538EA 109.18 USD
76774-538 76774-536 76774-534 76774-532 76774-544 76774-530 76774-542 76774-540
5-TAMRA 98%
5-TAMRA

Caution: for Research Use Only

Formula: C₂₅H₂₂N₂O₅
MW: 430.45 g/mol
MDL Number: MFCD00269769
CAS Number: 91809-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR