(6-(4-(tert-Butoxycarbonyl)piperazin-1-yl)pyridin-3-yl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A500470-250MG
A500470-1G
A500470-5G
A500470-100MG
76774-486EA
15
USD
76774-486
77878-277
77878-278
77878-276
(6-(4-(tert-Butoxycarbonyl)piperazin-1-yl)pyridin-3-yl)boronic acid 98%
(6-(4-(tert-Butoxycarbonyl)piperazin-1-yl)pyridin-3-yl)boronic acid
Caution: for Research Use Only
Formula:
C₁₄H₂₂BN₃O₄ |
MDL Number:
MFCD09952037 CAS Number: 919347-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...