4-Vinyl-1,3-dioxolan-2-one 98%

Supplier: Ambeed
Danger

A428917-5G A428917-25G A428917-100G
76773-902EA 6 USD
76773-902 77598-484 77878-181
4-Vinyl-1,3-dioxolan-2-one 98%
4-Vinyl-1,3-dioxolan-2-one

Caution: for Research Use Only

Formula: C₅H₆O₃
MW: 114.10 g/mol
MDL Number: MFCD00143315
CAS Number: 4427-96-7
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR