Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate 98%

Supplier: Ambeed
Danger

A373710-25G A373710-5G
76773-484EA 14 USD
76773-484 76773-486
Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate 98%
Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate

Caution: for Research Use Only

Formula: C₈H₁₁N₃O₃S
MDL Number: MFCD00071532
CAS Number: 64485-88-7
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR