Tributyrin (glycerol tributyrate) 98%

Supplier: Ambeed
Warning

A346664-100G A346664-25G
76773-244EA 20 USD
76773-244 77878-054
Tributyrin (glycerol tributyrate) 98%
Tributyrin (Glycerol tributyrate)

Caution: for Research Use Only

Formula: C₁₅H₂₆O₆
MDL Number: MFCD00009392
CAS Number: 60-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR