(2,3-Difluorophenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2,3-Difluorophenyl)acetic acid
A313966-25G
A313966-5G
A313966-100G
76772-824EA
34
USD
76772-824
76772-826
77877-999
(2,3-Difluorophenyl)acetic acid 98%
(2,3-Difluorophenyl)acetic acid
Caution: for Research Use Only
Formula:
(C₆H₃F₂)CH₂COOH MW: 172.13 g/mol |
MDL Number:
MFCD00040968 CAS Number: 145689-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...