3-(Trifluoromethyl)cinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(3-(Trifluoromethyl)phenyl)acrylic acid

A286061-100G A286061-10G A286061-25G A286061-500G A286061-5G
76772-390EA 61 USD
76772-390 76772-392 76772-394 76772-396 76772-398
3-(Trifluoromethyl)cinnamic acid 98%
3-(Trifluoromethyl)cinnamic acid

Caution: for Research Use Only

Formula: C₁₀H₇F₃O₂
MDL Number: MFCD00004393
CAS Number: 779-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR