Saccharin ≥98%

Supplier: Ambeed
Warning

A239294-100G A239294-10G A239294-1G A239294-500G A239294-25G A239294-1000G
76771-366EA 13 USD
76771-366 76771-368 76771-370 76771-372 77304-154 77550-544
Saccharin ≥98%
Saccharin

Caution: for Research Use Only

Formula: C₇H₅NO₃S
MDL Number: MFCD00005866
CAS Number: 81-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR