2,6-Dichlorobenzoxazole 98%

Supplier: Ambeed
Warning

A228495-100G A228495-10G A228495-1G A228495-500G A228495-5G A228495-25G
76771-066EA 26 USD
76771-066 76771-068 76771-070 76771-072 76771-074 77089-102
2,6-Dichlorobenzoxazole 98%
2,6-Dichlorobenzoxazole

Caution: for Research Use Only

Formula: C₇H₃Cl₂NO
MDL Number: MFCD07368635
CAS Number: 3621-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR