6-Amino-2-naphthalenesulfonic Acid 98% (by HPLC)

Supplier: Ambeed
Warning

A204871-100G A204871-5G A204871-25G
76770-288EA 30 USD
76770-288 76770-290 77626-038
6-Amino-2-naphthalenesulfonic Acid 98% (by HPLC)
6-Amino-2-naphthalenesulfonic Acid

Caution: for Research Use Only

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
MDL Number: MFCD00035736
CAS Number: 93-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR