1-Bromo-4-pentylbenzene 98%

Supplier: Ambeed
Warning

Synonyms: 4-Pentylbromobenzene, 4-Bromopentylbenzene, (4-Bromophenyl)pentane

A197105-1000G A197105-100G A197105-10G A197105-25G A197105-500G A197105-5G
76770-016EA 285 USD
76770-016 76770-018 76770-020 76770-022 76770-024 76770-026
1-Bromo-4-pentylbenzene 98%
1-Bromo-4-pentylbenzene

Caution: for Research Use Only

Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
MDL Number: MFCD00061113
CAS Number: 51554-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR