5-Iodopicolinic acid 97%

Supplier: Ambeed
Warning

A186931-100MG A186931-250MG A186931-5G A186931-1G
76769-776EA 27 USD
76769-776 76769-778 76769-780 77877-381
5-Iodopicolinic acid 97%
5-Iodopicolinic acid

Caution: for Research Use Only

Formula: C₆H₄INO₂
MDL Number: MFCD09910324
CAS Number: 32046-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR