5-Methyl-2-phenyl-2-hexenal 94% GC_ASSAY_METHOD

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-phenylhex-2-enal

A167660-100G A167660-10G A167660-25G A167660-5G
76768-946EA 85 USD
76768-946 76768-948 76768-950 76768-952
5-Methyl-2-phenyl-2-hexenal 94% GC_ASSAY_METHOD
5-Methyl-2-phenyl-2-hexenal

Caution: for Research Use Only

Formula: C₁₃H₁₆O
MW: 188.27 g/mol
MDL Number: MFCD00036615
CAS Number: 21834-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR