4-(Boc-amino)piperidine 97%

Supplier: Ambeed
Danger

Synonyms: 4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine

A157670-1000G A157670-10G A157670-25G A157670-500G A157670-5G A157670-100G
76768-432EA 517 USD
76768-432 76768-434 76768-436 76768-438 76768-440 77549-992
4-(Boc-amino)piperidine 97%
4-(Boc-amino)piperidine

Caution: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD00798171
CAS Number: 73874-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR